Dowex® 1X8 (Cl) 100-200 (Cl), beads

Supplier: BeanTown Chemical

133815-250G 133815-50G
BT133815-1KGEA 492.6 USD
BT133815-1KG BT133815-250G BT133815-50G
Dowex® 1X8 (Cl) 100-200 (Cl), beads
Dowex® 1X8 (Cl)

CAS: 69011-19-4; MDL No: MFCD00132718 Beads

MDL Number: MFCD00132718
CAS Number: 12627-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR