(1-Butyl)triphenylphosphonium bromide ≥99%
Supplier: BeanTown Chemical
Synonyms:
Butyltriphenylphosphonium bromide
133795-100G
133795-25G
133795-500G
BT133795-100GEA
83
USD
BT133795-100G
BT133795-25G
BT133795-500G
(1-Butyl)triphenylphosphonium bromide ≥99%
(1-Butyl)triphenylphosphonium bromide
CAS: 1779-51-7; EC No: 217-219-4; MDL No: MFCD00011855; RTECS: TA1855200
UN No: UN3464; Haz Class: 6.1; Packing Group: III
Crystalline; Linear Formula: (C6H5)3P(Br)CH2CH2CH2CH3; Molecular Formula: C22H24BrP; MW: 339.30
Melting Point: 240-243°
Hygroscopic
- RTECS: TA1855200
- Hygroscopic
- Crystalline
Formula:
H₃C(CH₂)₃P(C₆H₅)₃Br Melting Pt: 240…243 °C |
MDL Number:
MFCD00011855 CAS Number: 1779-51-7 EINECS: 217-219-4 UN: 3464 ADR: 6.1,III |
|
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...