2-Pyrimidinecarbonitrile ≥97%

Supplier: BeanTown Chemical

Synonyms: 2-Cyanopyrimidine

133650-25G 133650-5G
BT133650-25GEA 83 USD
BT133650-25G BT133650-5G
2-Pyrimidinecarbonitrile ≥97%
2-Pyrimidinecarbonitrile

CAS: 14080-23-0; MDL No: MFCD00160513 Solid; Molecular Formula: C5H3N3; MW: 105.10 Melting Point: 40-44°; Flash point: 110°C (230°F)


  • Solid

Formula: C₅H₃N₃
Melting Pt: 40…44 °C
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00160513
CAS Number: 14080-23-0

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR