Span® 40
Supplier: BeanTown Chemical
Synonyms:
Sorbitan monopalmitate, Arlacel 40
133545-250G
BT133545-250GEA
35.69
USD
BT133545-250G
Span® 40
Span® 40
CAS: 26266-57-9; EC No: 247-568-8; MDL No: MFCD00080946; RTECS: WG2932900
Crystalline; Molecular Formula: C22H42O6; MW: 402.57
Melting Point: 46-47°; Flash point: 113°C (235°F)
- RTECS: WG2932900
- Crystalline
Formula:
C₂₂H₄₂O₆ Melting Pt: 46…47 °C Flash Pt: 113 °C (235 °F) |
MDL Number:
MFCD00080946 CAS Number: 26266-57-9 EINECS: 247-568-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...