O-(tert-Butyldimethylsilyl)hydroxylamine ≥95%
Supplier: BeanTown Chemical
133535-250MG
BT133535-1GEA
87.72
USD
BT133535-1G
BT133535-250MG
O-(tert-Butyldimethylsilyl)hydroxylamine ≥95%
O-(tert-Butyldimethylsilyl)hydroxylamine
CAS: 41879-39-4; MDL No: MFCD00054944
UN No: UN3259; Haz Class: 8; Packing Group: III
Crystalline; Linear Formula: (CH3)3CSi(CH3)2ONH2; Molecular Formula: C6H17NOSi; MW: 147.29
Melting Point: 62-65°; Boiling Point: 87-90°/40 mmHg
Moisture Sensitive
- Moisture Sensitive
- Crystalline
Formula:
C₆H₁₇NOSi MW: 147.293 g/mol Boiling Pt: 87…90 °C/40 mmHg Melting Pt: 62…65 °C |
MDL Number:
MFCD00054944 CAS Number: 41879-39-4 ADR: 8 III |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...