4-Bromo-1-butene ≥97%
Supplier: BeanTown Chemical
133520-25G
133520-5G
BT133520-25GEA
234
USD
BT133520-25G
BT133520-5G
4-Bromo-1-butene ≥97%
4-Bromo-1-butene
CAS: 5162-44-7; EC No: 225-937-4; MDL No: MFCD00000258
UN No: UN1993; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: BrCH2CH2CH=CH2; Molecular Formula: C4H7Br; MW: 135.00
Boiling Point: 98-100°; Flash point: 1°C (34°F)
Density (g/mL): 1.33; Refractive Index: 1.462
Light Sensitive
- Refractive index: 1.462
- Light Sensitive
- Liquid
Formula:
C₄H₇Br Boiling Pt: 98…100 °C Density: 1.33 g/mL Flash Pt: 1 °C (34 °F) |
MDL Number:
MFCD00000258 CAS Number: 5162-44-7 EINECS: 225-937-4 UN: 1993 ADR: 3,II |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...