(S)-(+)-4-Phenyl-2-oxazolidinone ≥98%
Supplier: BeanTown Chemical
133445-1G
133445-5G
BT133445-1GEA
92
USD
BT133445-1G
BT133445-5G
(S)-(+)-4-Phenyl-2-oxazolidinone ≥98%
(S)-(+)-4-Phenyl-2-oxazolidinone
CAS: 99395-88-7; MDL No: MFCD00043396
Crystalline; Molecular Formula: C9H9NO2 ; MW: 163.17
Melting Point: 129-132°
Optical Rotation: [α]20/D +48°, c = 2 in chloroform
- Optical rotation: [α]20/D +48°, c = 2 in chloroform
- Crystalline
Formula:
C₉H₉NO₂ Melting Pt: 129…132 °C |
MDL Number:
MFCD00043396 CAS Number: 99395-88-7 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...