Titanium ≥99.95% (trace metals basis), wire Ø 0.75 mm

Supplier: BeanTown Chemical

133440-100CM 133440-25CM
BT133440-100CMEA 154.5 USD
BT133440-100CM BT133440-25CM
Titanium ≥99.95% (trace metals basis), wire Ø 0.75 mm
Titanium

CAS: 7440-32-6; EC No: 231-142-3; MDL No: MFCD00011264 Solid; Molecular Formula: Ti; MW: 47.87 Melting Point: 1660°; Boiling Point: 3287° Density (g/mL): 4.5


  • Solid

Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262
Melting Pt: 1660
Density: 4.5 g/mL
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now

Specification Test Results

Assay 99.95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR