Tetraammineplatinum(II) nitrate ≥99.99%
Supplier: BeanTown Chemical
133430-1G
133430-250MG
133430-5G
BT133430-1GEA
200.4
USD
BT133430-1G
BT133430-250MG
BT133430-5G
Tetraammineplatinum(II) nitrate ≥99.99%
Tetraammineplatinum(II) nitrate
CAS: 20634-12-2; EC No: 243-929-9; MDL No: MFCD00011624
UN No: UN1477; Haz Class: 5.1; Packing Group: III
Crystalline; Linear Formula: Pt(NH3)4(NO3)2; Molecular Formula: H12N6O6Pt; MW: 387.21
Melting Point: 262° (decomposes)
- Crystalline
Formula:
H₁₂N₆O₆Pt Melting Pt: 262 °C (dec.) |
MDL Number:
MFCD00011624 CAS Number: 20634-12-2 EINECS: 243-929-9 ADR: 5.1,III |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...