Calcium stearate

Supplier: BeanTown Chemical

Synonyms: Stearic acid calcium salt

133410-1KG 133410-5KG
BT133410-1KGEA 45.2 USD
BT133410-1KG BT133410-5KG
Calcium stearate
Calcium stearate

CAS: 1592-23-0; EC No: 216-472-8; MDL No: MFCD00036390 Powder; Linear Formula: Ca[O2C(CH2)16CH3]2; Molecular Formula: C26H70CaO4; MW: 607.04


  • Powder

Formula: C₃₆H₇₀CaO₄
MDL Number: MFCD00036390
CAS Number: 1592-23-0
EINECS: 216-472-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66064c431fc65dcdf98ff2bc1d0227f37de78108&vid=c1765b9467ae214d145f3824443319aa43d8bd1e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR