3-Bromo-o-toluidine ≥98%

Supplier: BeanTown Chemical

Synonyms: 2-Amino-6-bromotoluene, 3-Bromo-2-methylaniline

133310-100G 133310-25G 133310-5G
BT133310-100GEA 1055 USD
BT133310-100G BT133310-25G BT133310-5G
3-Bromo-o-toluidine ≥98%
3-Bromo-o-toluidine

CAS: 55289-36-6; EC No: 259-568-5; MDL No: MFCD00051579 UN No: UN2810; Haz Class: 6.1; Packing Group: III Solid; Molecular Formula: C7H8BrN; MW: 186.05 Boiling Point: 248°; Flash point: 110°C (230°F) Density (g/mL): 1.51; Refractive Index: 1.619 Light Sensitive


  • Refractive index: 1.619
  • Light Sensitive
  • Solid

Formula: C₇H₈BrN
Boiling Pt: 248 °C
Density: 1.51 g/mL
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00051579
CAS Number: 55289-36-6
EINECS: 259-568-5
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR