Pivaloyl chloride ≥98%

Supplier: BeanTown Chemical

Synonyms: Trimethylacetyl chloride

133270-100ML 133270-2.5L 133270-500ML
BT133270-100MLEA 42.13 USD
BT133270-100ML BT133270-2.5L BT133270-500ML
Pivaloyl chloride ≥98%
Pivaloyl chloride

CAS: 3282-30-2; EC No: 221-921-6; MDL No: MFCD00000709 UN No: UN2438; Haz Class: 6.1 (8,3); Packing Group: I Liquid; Linear Formula: (CH3)3CCOCl; Molecular Formula: C5H9ClO; MW: 120.58 Melting Point: -57°; Boiling Point: 105-106°; Flash point: 8°C (46°F) Density (g/mL): 0.979; Refractive Index: 1.412 Moisture Sensitive


  • Refractive index: 1.412
  • Moisture Sensitive
  • Liquid

Formula: C₅H₉ClO
MW: 120.58
Boiling Pt: 105
Melting Pt: -56
Density: 0.979 g/mL
Flash Pt: 8 °C (46 °F)
MDL Number: MFCD00000709
CAS Number: 3282-30-2
EINECS: 221-921-6
UN: 2438
ADR: 6.1 I

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR