Pivaloyl chloride ≥98%
Supplier: BeanTown Chemical
Synonyms:
Trimethylacetyl chloride
133270-100ML
133270-2.5L
133270-500ML
BT133270-100MLEA
42.13
USD
BT133270-100ML
BT133270-2.5L
BT133270-500ML
Pivaloyl chloride ≥98%
Pivaloyl chloride
CAS: 3282-30-2; EC No: 221-921-6; MDL No: MFCD00000709
UN No: UN2438; Haz Class: 6.1 (8,3); Packing Group: I
Liquid; Linear Formula: (CH3)3CCOCl; Molecular Formula: C5H9ClO; MW: 120.58
Melting Point: -57°; Boiling Point: 105-106°; Flash point: 8°C (46°F)
Density (g/mL): 0.979; Refractive Index: 1.412
Moisture Sensitive
- Refractive index: 1.412
- Moisture Sensitive
- Liquid
Formula:
C₅H₉ClO MW: 120.58 Boiling Pt: 105 Melting Pt: -56 Density: 0.979 g/mL Flash Pt: 8 °C (46 °F) |
MDL Number:
MFCD00000709 CAS Number: 3282-30-2 EINECS: 221-921-6 UN: 2438 ADR: 6.1 I |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...