2-Chloronicotinic acid ≥99%
Supplier: BeanTown Chemical
Synonyms:
2-Chloro-3-pyridinecarboxylic acid, 2-Chloropyridine-3-carboxylic acid
133225-10G
133225-250G
133225-50G
BT133225-10GEA
24
USD
BT133225-10G
BT133225-250G
BT133225-50G
2-Chloronicotinic acid ≥99%
2-Chloronicotinic acid
CAS: 2942-59-8; EC No: 220-937-0; MDL No: MFCD00006236
Powder; Molecular Formula: C6H4ClNO2; MW: 157.55
Melting Point: 176-178°
- Powder
Formula:
C₆H₄ClNO₂ Melting Pt: 176…178 °C |
MDL Number:
MFCD00006236 CAS Number: 2942-59-8 EINECS: 220-937-0 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...