2-Chloroethyl methyl ether ≥98%

Supplier: BeanTown Chemical

133215-100G 133215-25G 133215-500G
BT133215-100GEA 86 USD
BT133215-100G BT133215-25G BT133215-500G
2-Chloroethyl methyl ether ≥98%
2-Chloroethyl methyl ether

CAS: 627-42-9; EC No: 210-999-7; MDL No: MFCD00000972 UN No: UN3271; Haz Class: 3; Packing Group: II Liquid; Linear Formula: ClCH2CH2OCH3; Molecular Formula: C3H7ClO ; MW: 94.54 Boiling Point: 89-90°; Flash point: 10°C (50°F) Density (g/mL): 1.035; Refractive Index: 1.408


  • Refractive index: 1.408
  • Liquid

Formula: C₃H₇ClO
Boiling Pt: 89…90 °C
Density: 1.035 g/mL
Flash Pt: 10 °C (50 °F)
MDL Number: MFCD00000972
CAS Number: 627-42-9
EINECS: 210-999-7
UN: 3271
ADR: 3,II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR