Molecular sieve A4 (0.4 nm, 4 Å), beads 1 - 2 mm

Supplier: BeanTown Chemical

133105-1KG 133105-250G 133105-5KG
BT133105-1KGEA 124.29 USD
BT133105-1KG BT133105-250G BT133105-5KG
Molecular sieve A4 (0.4 nm, 4 Å), beads 1 - 2 mm
Molecular sieve A4 (0.4 nm, 4 Å)
MDL Number: MFCD03457537
CAS Number: 70955-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR