1-Bromo-3,5-dichlorobenzene ≥97%

Supplier: BeanTown Chemical

Synonyms: 3,5-Dichlorobromobenzene

133040-100G 133040-25G 133040-5G
BT133040-100GEA 211 USD
BT133040-100G BT133040-25G BT133040-5G
1-Bromo-3,5-dichlorobenzene ≥97%
1-Bromo-3,5-dichlorobenzene

CAS: 19752-55-7; EC No: 243-270-7; MDL No: MFCD00000584 Powder; Molecular Formula: C6H3BrCl2 ; MW: 225.90 Melting Point: 73-75°; Boiling Point: 232°


  • Powder

Formula: C₆H₃BrCl₂
Boiling Pt: 232 °C
Melting Pt: 73…75 °C
MDL Number: MFCD00000584
CAS Number: 19752-55-7
EINECS: 243-270-7

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR