3,5-Dichloro-4-methylpyridine

Supplier: BeanTown Chemical

133035-1G 133035-250MG
BT133035-1GEA 318 USD
BT133035-1G BT133035-250MG
3,5-Dichloro-4-methylpyridine
3,5-Dichloro-4-methylpyridine

CAS: 100868-46-0 Solid; Molecular Formula: C6H5Cl2N; MW: 162.02


  • Solid

Formula: C₆H₅Cl₂N
MW: 162.013 g/mol
MDL Number: MFCD09839290
CAS Number: 100868-46-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR