Holmium(III) oxide ≥99.9%
Supplier: BeanTown Chemical
Synonyms:
Diholmium trioxide, Holmium oxide
132945-10G
132945-50G
BT132945-10GEA
170
USD
BT132945-10G
BT132945-50G
Holmium(III) oxide ≥99.9%
Holmium (III) oxide
CAS: 12055-62-8; EC No: 235-015-3; MDL No: MFCD00011053
3-12mm Sintered Lump; Molecular Formula: Ho2O3; MW: 377.86
Melting Point: 2330°
- 3-12mm Sintered Lump
Formula:
Ho₂O₃ Melting Pt: 2330 °C |
MDL Number:
MFCD00011053 CAS Number: 12055-62-8 EINECS: 235-015-3 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...