2-Bromo-3-nitrobenzoic acid ≥90%, Technical Grade

Supplier: BeanTown Chemical

132930-1G 132930-250MG
BT132930-1GEA 66 USD
BT132930-1G BT132930-250MG
2-Bromo-3-nitrobenzoic acid ≥90%, Technical Grade
2-Bromo-3-nitrobenzoic acid

CAS: 573-54-6; EC No: 209-356-3; MDL No: MFCD00074899 Crystalline/Powder; Molecular Formula: C7H4BrNO4 ; MW: 246.01 Melting Point: 184-186°


  • Crystalline/Powder

Formula: C₇H₄BrNO₄
MW: 246.02 g/mol
Melting Pt: 184…186 °C
MDL Number: MFCD00074899
CAS Number: 573-54-6
EINECS: 209-356-3

Order Now

Specification Test Results

Assay 90%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR