Bis(2-ethylhexyl)phosphate ≥95%
Supplier: BeanTown Chemical
Synonyms:
Bis(2-ethylhexyl) hydrogen phosphate, Di-(2-ethylhexyl)phosphoric acid
132885-100G
132885-2KG
132885-500G
BT132885-100GEA
29
USD
BT132885-100G
BT132885-2KG
BT132885-500G
Bis(2-ethylhexyl)phosphate ≥95%
Bis(2-ethylhexyl) phosphate
CAS: 298-07-7; EC No: 206-056-4; MDL No: MFCD00009492; RTECS: TB7875000
UN No: UN1902; Haz Class: 8; Packing Group: III
Liquid; Molecular Formula: C16H35O4P; MW: 322.42
Melting Point: -60°; Boiling Point: 48°/12 mmHg; Flash point: 130°C (266°F)
Density (g/mL): 0.965; Refractive Index: 1.443
- RTECS: TB7875000
- Refractive index: 1.443
- Liquid
Formula:
C₁₆H₃₅O₄P Boiling Pt: 48 °C/12 mmHg Melting Pt: –60 °C Density: 0.965 g/mL Flash Pt: 130 °C (266 °F) |
MDL Number:
MFCD00009492 CAS Number: 298-07-7 EINECS: 206-056-4 UN: 1902 ADR: 8,III |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...