Methyl-2-amino-5-bromobenzoate ≥98%
Supplier: BeanTown Chemical
132815-100G
132815-25G
132815-5G
BT132815-100GEA
341
USD
BT132815-100G
BT132815-25G
BT132815-5G
Methyl-2-amino-5-bromobenzoate ≥98%
Methyl 2-amino-5-bromobenzoate
CAS: 52727-57-8; EC No: 000-000-0; MDL No: MFCD00144761
Solid; Molecular Formula: C8H8BrNO2; MW: 230.06
Melting Point: 72-74°
- Solid
Formula:
C₈H₈BrNO₂ Melting Pt: 72…74 °C |
MDL Number:
MFCD00144761 CAS Number: 52727-57-8 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...