Niobium ≥99.5% (trace metals basis), foil, Thickness 0.025 mm
Supplier: BeanTown Chemical
132795-20X100
132795-20X20CM
BT132795-20X10EA
317.42
USD
BT132795-20X10
BT132795-20X20
Niobium ≥99.5% (trace metals basis), foil, Thickness 0.025 mm
Niobium
CAS: 7440-03-1; EC No: 231-113-5; MDL No: MFCD00011126
Powder; Molecular Formula: Nb ; MW: 92.91
Melting Point: 2468°; Boiling Point: 4742°
Density (g/mL): 8.57
- Powder
Formula:
Nb MW: 92.91 g/mol Boiling Pt: 3700 Melting Pt: 2468 Density: 8.57 g/mL |
MDL Number:
MFCD00011126 CAS Number: 7440-03-1 EINECS: 231-113-5 |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...