Dimethyl sulfoxide ≥99%

Supplier: BeanTown Chemical

Synonyms: Methanesulphinylmethane, Methyl sulfoxide, Dimethyl(oxido)sulfur, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO, Methanesulfinylmethane

132780-10KG 132780-2.5KG 132780-500G
BT132780-10KGEA 238.24 USD
BT132780-10KG BT132780-2.5KG BT132780-500G
Dimethyl sulfoxide ≥99%
Dimethyl sulphoxide

CAS: 67-68-5; EC No: 200-664-3; MDL No: MFCD00002089; RTECS: PV6210000 Liquid; Linear Formula: (CH3)2SO ; MW: 78.13 Melting Point: 16-19°; Boiling Point: 189°; Flash point: 87°C (189°F) Density (g/mL): 1.101; Refractive Index: 1.479 Hygroscopic


  • RTECS: PV6210000
  • Refractive index: 1.479
  • Hygroscopic
  • Liquid

Formula: (CH₃)₂SO
MW: 78.14 g/mol
Boiling Pt: 189
Melting Pt: 18.5
Density: 1.101 g/mL
Flash Pt: 87 °C (189 °F)
MDL Number: MFCD00002089
CAS Number: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR