1-(2-Pyridylazo)-2-naphthol (PAN) ≥98%

Supplier: BeanTown Chemical

Synonyms: PAN, 1-(2-Pyridylazo)-2-naphthol

132775-1G 132775-25G 132775-5G
BT132775-1GEA 21 USD
BT132775-1G BT132775-25G BT132775-5G
1-(2-Pyridylazo)-2-naphthol (PAN) ≥98%
1-(2-Pyridylazo)-2-naphthol

CAS: 85-85-8; EC No: 201-637-9; MDL No: MFCD00004071 Crystalline; Molecular Formula: C15H11N3O; MW: 249.27 Melting Point: 138-141°


  • Crystalline

Formula: C₁₅H₁₁N₃O
MW: 249.27 g/mol
Boiling Pt: 412
Melting Pt: 138…141 °C
MDL Number: MFCD00004071
CAS Number: 85-85-8
EINECS: 201-637-9

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR