D-(+)-Glucose monohydrate ≥99%

Supplier: BeanTown Chemical

Synonyms: (2S,3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol hydrate, Dextrose monohydrate

132735-2.5KG 132735-500G
BT132735-2.5KGEA 103.24 USD
BT132735-2.5KG BT132735-500G
D-(+)-Glucose monohydrate ≥99%
D(+)-Glucose monohydrate

CAS: 14431-43-7; EC No: 200-075-1; MDL No: MFCD00149450 Powder; Molecular Formula: C6H12O6·H2O; MW: 198.18


  • Powder

Formula: C₆H₁₂O₆·H₂O
MW: 198.17 g/mol
Melting Pt: 83
MDL Number: MFCD00149450
CAS Number: 14431-43-7
EINECS: 200-075-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9ea2af2ca62ca94d86809070593a60aafd8cd1b&vid=e549b499dca7fb95811640efad1429268c8f6d8b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR