Cerium trichloride heptahydrate ≥99%

Supplier: BeanTown Chemical

Synonyms: Cerous chloride heptahydrate, Cerium (III) chloride heptahydrate

132700-1KG 132700-250G 132700-50G
BT132700-1KGEA 339 USD
BT132700-1KG BT132700-250G BT132700-50G
Cerium trichloride heptahydrate ≥99%
Cerium trichloride heptahydrate

CAS: 18618-55-8; EC No: 232-227-8; MDL No: MFCD00149634; RTECS: FK5100000 Crystalline; Molecular Formula: CeCl3·7H2O; MW: 372.58 Density (g/mL): 3.94 Air Sensitive, Hygroscopic


  • RTECS: FK5100000
  • Air Sensitive, Hygroscopic
  • Crystalline

Formula: CeCl₃·7H₂O
Density: 3.94 g/mL
MDL Number: MFCD00149634
CAS Number: 18618-55-8
EINECS: 232-227-8

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR