2-Chloro-5-methoxybenzimidazole ≥94%

Supplier: BeanTown Chemical

Synonyms: 2-Chloro-5-methoxy-1H-benzo[d]imidazole, 2-Chloro-5-methoxy-1H-benzimidazole

132685-1G 132685-250MG
BT132685-1GEA 211 USD
BT132685-1G BT132685-250MG
2-Chloro-5-methoxybenzimidazole ≥94%
2-Chloro-5-methoxybenzimidazole

CAS: 15965-54-5; MDL No: MFCD00051945 Powder; Molecular Formula: C8H7ClN2O; MW: 182.61


  • Powder

Formula: C₈H₇ClN₂O
MDL Number: MFCD00051945
CAS Number: 15965-54-5

Order Now

Specification Test Results

Assay 94%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR