Sulfanilic acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
Aniline-4-sulphonic acid, 4-Aminobenzenesulphonic acid, 4-Aminobenzenesulfonic acid, Aniline-4-sulfonic acid
132640-1KG
132640-250G
132640-5KG
BT132640-1KGEA
57.08
USD
BT132640-1KG
BT132640-250G
BT132640-5KG
Sulfanilic acid ≥98%
Sulphanilic acid
CAS: 121-57-3; EC No: 204-482-5; MDL No: MFCD00007886; RTECS: WP3895500
Crystalline/Pieces; Linear Formula: H2NC6H4SO3H; Molecular Formula: C6H7NO3S; MW: 173.19
Melting Point: 365°
Density (g/mL): 1.485
- RTECS: WP3895500
- Crystalline/Pieces
Formula:
4-(H₂N)C₆H₄SO₃H MW: 173.19 g/mol Boiling Pt: 363 Melting Pt: 279 Density: 1.485 g/mL |
MDL Number:
MFCD00007886 CAS Number: 121-57-3 EINECS: 204-482-5 Merck Index: 13,09012 |
|
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...