Boranepyridine complex ≥95%
Supplier: BeanTown Chemical
Synonyms:
Pyridine Borane, Borane, pyridine complex
132600-10G
BT132600-10GEA
58
USD
BT132600-10G
Boranepyridine complex ≥95%
Boranepyridine complex
CAS: 110-51-0; EC No: 203-773-4; MDL No: MFCD00012435; RTECS: US3675000
UN No: UN2810; Haz Class: 6.1; Packing Group: II
Liquid; Molecular Formula: C5H8BN; MW: 92.93
Melting Point: 10-11°; Flash point: 21°C (70°F)
Density (g/mL): 0.92; Refractive Index: 1.532
Air Sensitive, Moisture Sensitive
- RTECS: US3675000
- Refractive index: 1.532
- Air Sensitive, Moisture Sensitive
- Liquid
Formula:
C₅H₈BN Melting Pt: 10…11 °C Density: 0.92 g/mL Flash Pt: 21 °C (70 °F) |
MDL Number:
MFCD00012435 CAS Number: 110-51-0 EINECS: 203-773-4 ADR: 6.1,II |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...