Zinc ferrite

Supplier: BeanTown Chemical

132465-100G 132465-25G
BT132465-100GEA 369 USD
BT132465-100G BT132465-25G
Zinc ferrite
Zinc ferrite

CAS: 12063-19-3; EC No: 235-052-5; MDL No: MFCD00049621 Powder; Linear Formula: ZnFe2O4; MW: 241.08


  • Powder

Formula: Fe₂H₂O₄Zn
MDL Number: MFCD00049621
CAS Number: 12063-19-3
EINECS: 235-052-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR