α-Bromo-4-chlorotoulene ≥98%

Supplier: BeanTown Chemical

Synonyms: alpha-Bromo-4-chlorotoluene, 1-(Bromomethyl)-4-chlorobenzene, 4-Chlorobenzyl bromide

132385-100G 132385-25G 132385-5G
BT132385-100GEA 558 USD
BT132385-100G BT132385-25G BT132385-5G
α-Bromo-4-chlorotoulene ≥98%
α-Bromo-4-chlorotoulene

CAS: 622-95-7; EC No: 210-760-7; MDL No: MFCD00040714 UN No: UN3261; Haz Class: 8; Packing Group: II Solid; Molecular Formula: C7H6BrCl; MW: 205.48 Melting Point: 48-52°


  • Solid

Formula: C₇H₆BrCl
MW: 205.48 g/mol
Melting Pt: 48…52 °C
MDL Number: MFCD00040714
CAS Number: 622-95-7
EINECS: 210-760-7
UN: 3261
ADR: 8 II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR