Copper(I) iodide ≥99.995%
Supplier: BeanTown Chemical
Synonyms:
Cuprous iodide
132300-100G
132300-25G
132300-5G
BT132300-100GEA
392
USD
BT132300-100G
BT132300-25G
BT132300-5G
Copper(I) iodide ≥99.995%
Copper (I) iodide
CAS: 7681-65-4; EC No: 231-674-6; MDL No: MFCD00010978
UN No: UN3077; Haz Class: 9; Packing Group: III
Powder/Chunks; Molecular Formula: CuI; MW: 190.45
Melting Point: 605°
Density (g/mL): 5.62
Air Sensitive, Light Sensitive, Moisture Sensitive
- Air Sensitive, Light Sensitive, Moisture Sensitive
- Powder/Chunks
Formula:
CuI Melting Pt: 605 °C Density: 5.62 g/mL |
MDL Number:
MFCD00010978 CAS Number: 7681-65-4 EINECS: 231-674-6 UN: 3077 ADR: 9,III |
Specification Test Results
Assay | 99.995% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...