Lanthanum ≥99.9% (trace rare earth metals basis), powder -40 mesh
Supplier: BeanTown Chemical
132250-25G
132250-5G
BT132250-25GEA
268
USD
BT132250-25G
BT132250-5G
Lanthanum ≥99.9% (trace rare earth metals basis), powder -40 mesh
Lanthanum
CAS: 7439-91-0; EC No: 231-099-0; MDL No: MFCD00011066
UN No: UN3089; Haz Class: 4.1; Packing Group: II
Powder, ampuled under argon; Molecular Formula: La; MW: 138.91
Melting Point: 920°; Boiling Point: 3464°
Density (g/mL): 6.19
- Powder, ampuled under argon
Formula:
La Boiling Pt: 3464 °C Melting Pt: 920 °C Density: 6.19 g/mL |
MDL Number:
MFCD00011066 CAS Number: 7439-91-0 EINECS: 231-099-0 ADR: 4.1,II |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...