Lanthanum ≥99.9% (trace rare earth metals basis), powder -40 mesh

Supplier: BeanTown Chemical

132250-25G 132250-5G
BT132250-25GEA 268 USD
BT132250-25G BT132250-5G
Lanthanum ≥99.9% (trace rare earth metals basis), powder -40 mesh
Lanthanum

CAS: 7439-91-0; EC No: 231-099-0; MDL No: MFCD00011066 UN No: UN3089; Haz Class: 4.1; Packing Group: II Powder, ampuled under argon; Molecular Formula: La; MW: 138.91 Melting Point: 920°; Boiling Point: 3464° Density (g/mL): 6.19


  • Powder, ampuled under argon

Formula: La
Boiling Pt: 3464 °C
Melting Pt: 920 °C
Density: 6.19 g/mL
MDL Number: MFCD00011066
CAS Number: 7439-91-0
EINECS: 231-099-0
ADR: 4.1,II

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR