Palladium(II) trifluoroacetate ≥97%

Supplier: BeanTown Chemical

Synonyms: Palladium(II) 2,2,2-trifluoroacetate

132180-1G 132180-5G
BT132180-1GEA 113 USD
BT132180-1G BT132180-5G
Palladium(II) trifluoroacetate ≥97%
Palladium (II) trifluoroacetate

CAS: 42196-31-6; MDL No: MFCD00013204 Powder; Linear Formula: Pd(OOCCF3)2; MW: 332.44 Melting Point: 220° (decomposes) Hygroscopic


  • Hygroscopic
  • Powder

Formula: C₄F₆O₄Pd
MW: 332.45 g/mol
Melting Pt: 220 °C (dec.)
MDL Number: MFCD00013204
CAS Number: 42196-31-6

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR