4-Bromomethyl-2-cyanobiphenyl ≥98%

Supplier: BeanTown Chemical

Synonyms: 4'-(Bromomethyl)-[1,1'-biphenyl]-2-carbonitrile

132145-1G 132145-25G 132145-5G
BT132145-1GEA 17 USD
BT132145-1G BT132145-25G BT132145-5G
4-Bromomethyl-2-cyanobiphenyl ≥98%
4-Bromomethyl-2-cyanobiphenyl

CAS: 114772-54-2; EC No: 425-280-5; MDL No: MFCD00671503 UN No: UN3439; Haz Class: 6.1; Packing Group: III Powder; Molecular Formula: C14H10BrN; MW: 272.14 Melting Point: 125-128°


  • Powder

Formula: C₁₄H₁₀BrN
MW: 272.14 g/mol
Melting Pt: 125…128 °C
MDL Number: MFCD00671503
CAS Number: 114772-54-2
EINECS: 425-280-5
ADR: 6.1 III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR