Dipotassium hexachloroiridate

Supplier: BeanTown Chemical

Synonyms: Potassium hexachloroiridate(IV), Potassium hexachloroiridate (IV)

132140-1G 132140-25G 132140-5G
BT132140-1GEA 105 USD
BT132140-1G BT132140-25G BT132140-5G
Dipotassium hexachloroiridate
Dipotassium hexachloroiridate

CAS: 16920-56-2; EC No: 240-976-7; MDL No: MFCD00011369 Crystalline/Powder; Linear Formula: K2IrCl6; MW: 483.12 Melting Point: 385° Hygroscopic


  • Hygroscopic
  • Crystalline/Powder

Formula: Cl₆IrK₂
Melting Pt: >385 °C
MDL Number: MFCD00011369
CAS Number: 16920-56-2
EINECS: 240-976-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR