m-Cresol purple

Supplier: BeanTown Chemical

Synonyms: m-Cresolsulfonphthalein, 2',2''-Dimethylphenolsulphonphthalein, m-Cresolsulphonphthalein, MCP, 3,3-Bis(4-hydroxy-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, 2',2''-Dimethylphenolsulfonphthalein

132025-25G 132025-5G
BT132025-25GEA 183 USD
BT132025-25G BT132025-5G
m-Cresol purple
m-Cresol purple

CAS: 2303-01-7; EC No: 218-960-6; MDL No: MFCD00005871 Powder; Molecular Formula: C21H18O5S; MW: 382.44


  • Powder

Formula: C₂₁H₁₈O₅S
MW: 382.43 g/mol
MDL Number: MFCD00005871
CAS Number: 2303-01-7
EINECS: 218-960-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR