D-(-)-Arabinose ≥99%

Supplier: BeanTown Chemical

Synonyms: D-(-)-Arabinose, (2S,3R,4R)-2,3,4,5-Tetrahydroxypentanal

131985-10G 131985-250G 131985-50G
BT131985-10GEA 37.55 USD
BT131985-10G BT131985-250G BT131985-50G
D-(-)-Arabinose ≥99%
D(-)-Arabinose

CAS: 10323-20-3; EC No: 233-708-5; MDL No: MFCD00135608 Powder; Molecular Formula: C5H10O5; MW: 150.13 Melting Point: 162-164°


  • Powder

Formula: C₅H₁₀O₅
Melting Pt: 162…164 °C
MDL Number: MFCD00135608
CAS Number: 10323-20-3
EINECS: 233-708-5

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR