Allylboronic acid pinacol ester ≥98%

Supplier: BeanTown Chemical

Synonyms: 2-Allyl-4,4,5,5-Tetramethyl-1,3,2-Dioxaborolane

131900-1G 131900-5G
BT131900-1GEA 38 USD
BT131900-1G BT131900-5G
Allylboronic acid pinacol ester ≥98%
Allylboronic acid pinacol ester

CAS: 72824-04-5; MDL No: MFCD00013347 UN No: UN3272; Haz Class: 3; Packing Group: III Liquid; Molecular Formula: C9H17BO2; MW: 168.04 Boiling Point: 50-53°/5 mmHg; Flash point: 46°C (115°F) Density (g/mL): 0.896; Refractive Index: 1.427


  • Refractive index: 1.427
  • Liquid

Formula: C₉H₁₇BO₂
MW: 168.05 g/mol
Boiling Pt: 50…53 °C/5 mmHg
Density: 0.896 g/mL
Flash Pt: 46 °C (115 °F)
MDL Number: MFCD00013347
CAS Number: 72824-04-5
UN: 3272
ADR: 3 III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR