(R)-(+)-1-(1-Naphthyl)ethylamine ≥98%, ee 98%

Supplier: BeanTown Chemical

Synonyms: (R)-(+)-α-(1-Naphthyl)ethylamine, (R)-1-(Naphthalen-1-yl)ethanamine, (+)-1-(1-Naphthyl)ethylamine

131785-1G 131785-25G 131785-5G
BT131785-1GEA 62.97 USD
BT131785-1G BT131785-25G BT131785-5G
(R)-(+)-1-(1-Naphthyl)ethylamine ≥98%, ee 98%
(R)-(+)-1-(1-Naphthyl)ethylamine

CAS: 3886-70-2; EC No: 223-425-5; MDL No: MFCD00064114 UN No: UN2922; Haz Class: 8 (6.1); Packing Group: III Liquid; Molecular Formula: C12H13N; MW: 171.24 Boiling Point: 153°/11 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.067; Refractive Index: 1.623; Optical Rotation: [α]20/D +55°, c = 2 in ethanol Air Sensitive


  • Optical rotation: [α]20/D +55°, c = 2 in ethanol
  • Refractive index: 1.623
  • Air Sensitive
  • Liquid

Formula: C₁₂H₁₃N
Boiling Pt: 153 °C/11 mmHg
Density: 1.067 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00064114
CAS Number: 3886-70-2
EINECS: 223-425-5
ADR: 8 (6.1),III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR