1,1,3,3-Tetramethylguanidine ≥99%

Supplier: BeanTown Chemical

Synonyms: N,N,N',N'-Tetramethylguanidine

131625-100ML 131625-25ML 131625-500ML
BT131625-100MLEA 56.54 USD
BT131625-100ML BT131625-25ML BT131625-500ML
1,1,3,3-Tetramethylguanidine ≥99%
1,1,3,3-Tetramethylguanidine

CAS: 80-70-6; EC No: 201-302-7; MDL No: MFCD00008323 UN No: UN2920; Haz Class: 8 (3); Packing Group: II Liquid; Molecular Formula: C5H13N3; MW: 115.18 Boiling Point: 52-54°/11 mmHg; Flash point: 60°C (140°F) Density (g/mL): 0.918; Refractive Index: 1.407 Air Sensitive


  • Refractive index: 1.407
  • Air Sensitive
  • Liquid

Formula: C₅H₁₃N₃
Boiling Pt: 52…54 °C/11 mmHg
Density: 0.918 g/mL
Flash Pt: 60 °C (140 °F)
MDL Number: MFCD00008323
CAS Number: 80-70-6
EINECS: 201-302-7
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR