N,N-Diethyl-p-phenylenediammonium sulfate ≥97%
Supplier: BeanTown Chemical
Synonyms:
4-Amino-N,N-diethylaniline sulfate, N,N-Diethyl-p-phenylenediamine sulphate, N,N-Diethyl-1,4-phenylenediammonium sulphate, N1,N1-Diethylbenzene-1,4-diamine sulfate, 4-Amino-N,N-diethylaniline sulphate, N,N-Diethyl-1,4-phenylenediammonium sulfate, N,N-Diethyl-p-phenylenediamine sulfate, N,N-Diethylbenzene-1,4-diammonium sulfate, N1,N1-Diethylbenzene-1,4-diamine sulphate, N,N-Diethylbenzene-1,4-diammonium sulphate
CAS: 6283-63-2; EC No: 228-500-6; MDL No: MFCD00012993; RTECS: SS9625000
Solid; Linear Formula: (CH3CH2)2NC6H4NH2·H2SO4; Molecular Formula: C10H16N2·H2O4S; MW: 262.33
Melting Point: 184-186°
Air Sensitive, Light Sensitive
- RTECS: SS9625000
- Air Sensitive, Light Sensitive
- Solid
Formula:
C₁₀H₁₈N₂O₄S MW: 262.33 g/mol Melting Pt: 184…186 °C |
MDL Number:
MFCD00012993 CAS Number: 6283-63-2 EINECS: 228-500-6 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...