Diphenylcyclopropenone, (max. 5% H₂O) ≥99% (dry weight)

Supplier: BeanTown Chemical

Synonyms: 2,3-Diphenyl-2-cyclopropen-1-one, 2,3-Diphenylcycloprop-2-en-1-one, 2,3-Diphenyl-2-cyclopropen-1-on, 1,2-Diphenylcyclopropen-3-one

131580-1G 131580-25G 131580-5G
BT131580-1GEA 62 USD
BT131580-1G BT131580-25G BT131580-5G
Diphenylcyclopropenone, (max. 5% H₂O) ≥99% (dry weight)
Diphenylcyclopropenone

CAS: 886-38-4; EC No: 212-948-4; MDL No: MFCD00001311 Powder; Molecular Formula: C15H10O; MW: 206.24 Melting Point: 118-122°


  • Powder

Formula: C₁₅H₁₀O
MW: 206.24 g/mol
Boiling Pt: 407
Melting Pt: 118…122 °C
MDL Number: MFCD00001311
CAS Number: 886-38-4
EINECS: 212-948-4
Merck Index: 12,03366

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR