Calcium sulfate, Drierite™ 10-20 mesh for drying, with indicator

Supplier: BeanTown Chemical

Synonyms: Calcium sulphate (anhydrous) anhydrite, Gypsum, Calcium sulfate (anhydrous) anhydrite

131570-1LB 131570-5LB
BT131570-1LBEA 70.86 USD
BT131570-1LB BT131570-5LB
Calcium sulfate, Drierite™ 10-20 mesh for drying, with indicator
Calcium sulphate

CAS: 7778-18-9; EC No: 231-900-3; MDL No: MFCD00010912 Granular Moisture Sensitive


  • Moisture Sensitive
  • Granular

Formula: CaO₄S
MW: 136.14 g/mol
Melting Pt: 1350
MDL Number: MFCD00010912
CAS Number: 7778-18-9
EINECS: 231-900-3
Merck Index: 13,01711

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR