5-Fluorouracil ≥99%
Supplier: BeanTown Chemical
Synonyms:
Fluorouracil, 5-Fluoropyrimidine-2,4(1H,3H)-dione, 5-FU
131560-100G
131560-25G
131560-5G
BT131560-100GEA
1031
USD
BT131560-100G
BT131560-25G
BT131560-5G
5-Fluorouracil ≥99%
5-Fluorouracil
CAS: 51-21-8; EC No: 200-085-6; MDL No: MFCD00006018; RTECS: YR0350000
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Powder; Molecular Formula: C4H3FN2O2; MW: 130.08
Melting Point: 282-286° (decomposes)
Light Sensitive
- RTECS: YR0350000
- Light Sensitive
- Powder
Formula:
C₄H₃FN₂O₂ Melting Pt: 282…286 °C (dec.) |
MDL Number:
MFCD00006018 CAS Number: 51-21-8 EINECS: 200-085-6 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...