Ethyl oleate ≥70%, Technical Grade

Supplier: BeanTown Chemical

Synonyms: Elaidic acid ethyl ester, (Z)-9-Octadecenoic acid ethyl ester, Oleic acid ethyl ester

131540-1KG 131540-250G 131540-5KG
BT131540-1KGEA 97 USD
BT131540-1KG BT131540-250G BT131540-5KG
Ethyl oleate ≥70%, Technical Grade
Ethyl oleate

CAS: 111-62-6; EC No: 203-889-5; MDL No: MFCD00009579; RTECS: RG3715000 Liquid; Molecular Formula: C20H38O2; MW: 310.51 Melting Point: -32°; Boiling Point: 205-208°; Flash point: 110°C (230°F) Density (g/mL): 0.87; Refractive Index: 1.451 Light Sensitive


  • RTECS: RG3715000
  • Refractive index: 1.451
  • Light Sensitive
  • Liquid

Formula: C₂₀H₃₈O₂
Boiling Pt: 205…208 °C
Melting Pt: –32 °C
Density: 0.87 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00009579
CAS Number: 111-62-6
EINECS: 203-889-5

Order Now

Specification Test Results

Assay 70%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR