Chromium ≥99.95%, cubes 6.35 mm square
Supplier: BeanTown Chemical
131520-10G
131520-250G
131520-50G
BT131520-10GEA
40
USD
BT131520-10G
BT131520-250G
BT131520-50G
Chromium ≥99.95%, cubes 6.35 mm square
Chromium
CAS: 7440-47-3; EC No: 231-157-5; MDL No: MFCD00010944
Solid; Molecular Formula: Cr; MW: 52.00
Melting Point: 1857°; Boiling Point: 2672°
Density (g/mL): 7.14
- Solid
Formula:
Cr Boiling Pt: 2672 °C Melting Pt: 1857 °C Density: 7.14 g/mL |
MDL Number:
MFCD00010944 CAS Number: 7440-47-3 EINECS: 231-157-5 |
Specification Test Results
Assay | 99.95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...