Calcium carbonate ≥99.99% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Marble
131495-100G
131495-25G
131495-5G
BT131495-100GEA
407
USD
BT131495-100G
BT131495-25G
BT131495-5G
Calcium carbonate ≥99.99% (trace metals basis)
Calcium carbonate
CAS: 471-34-1; EC No: 207-439-9; MDL No: MFCD00010906; RTECS: FF9335000
Powder; Linear Formula: CaCO3; MW: 100.09
Density (g/mL): 2.93
- RTECS: FF9335000
- Powder
Formula:
CaCO₃ Density: 2.93 g/mL |
MDL Number:
MFCD00010906 CAS Number: 471-34-1 EINECS: 207-439-9 |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...